accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 54454-10-3

CH3NNCH3Cl

CAS RN 54454-10-3

IUPAC Name: 5-chloro-1,3-dimethyl-1H-pyrazole
Synonyms: 1,3-dimethyl-5-chloropyrazole
Molecular Weight (g/mol): 130.58
Molecular Formula: C5H7ClN2
InChi Key: DDUSLFAWARYAPR-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)