accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 54451-24-0

OOOOOOOOOLaLa

CAS RN 54451-24-0

IUPAC Name: dilanthanum(3+) hydrate tricarbonate
Synonyms: lanthanum carbonate
Molecular Weight (g/mol): 475.85
Molecular Formula: C3H2La2O10
InChi Key: AFCUGQOTNCVYSW-UHFFFAOYSA-H
SMILES: O.[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)