accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HO

CAS RN 5445-30-7

IUPAC Name: 1-butylcyclohexan-1-ol
Synonyms: 1-butylcyclohexan-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: RCHLXMOXBJRGNX-UHFFFAOYSA-N
SMILES: CCCCC1(O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results