accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 544-00-3

H3CCH3HNCH3CH3

CAS RN 544-00-3

IUPAC Name: bis(3-methylbutyl)amine
Synonyms: bis(3-methylbutyl)amine
Molecular Weight (g/mol): 157.3
Molecular Formula: C10H23N
InChi Key: SPVVMXMTSODFPU-UHFFFAOYSA-N
SMILES: CC(C)CCNCCC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results