accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SrCH3OOCH3OO

CAS RN 543-94-2

IUPAC Name: strontium(2+) diacetate
Synonyms: strontium acetate
Molecular Weight (g/mol): 205.71
Molecular Formula: C4H6O4Sr
InChi Key: RXSHXLOMRZJCLB-UHFFFAOYSA-L
SMILES: [Sr++].CC([O-])=O.CC([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results