accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5428-41-1

ONHNOHN

CAS RN 5428-41-1

IUPAC Name: 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
Synonyms: 5-cyanouracil
Molecular Weight (g/mol): 137.1
Molecular Formula: C5H3N3O2
InChi Key: HAUXRJCZDHHADG-UHFFFAOYSA-N
SMILES: O=C1NC=C(C#N)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results