accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHONHOHNO

CAS RN 5427-26-9

IUPAC Name: 2-(2,5-dioxoimidazolidin-4-yl)acetic acid
Synonyms: (2,5-dioxoimidazolidin-4-yl)acetic acid
Molecular Weight (g/mol): 158.11
Molecular Formula: C5H6N2O4
InChi Key: DQQLZADYSWBCOX-UHFFFAOYNA-N
SMILES: OC(=O)CC1NC(=O)NC1=O
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results