CAS RN 5426-62-0
CAS RN 5426-62-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Benzyl-Beta-alanine, TRC
CAS: 5426-62-0 Molecular Formula: C10H13NO2 Molecular Weight (g/mol): 179.22 IUPAC Name: 3-(benzylamino)propanoic acid SMILES: OC(=O)CCNCc1ccccc1