accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5423-07-4

OHNNNOONOOSOOOSOOONaNa(E)

CAS RN 5423-07-4

IUPAC Name: disodium 3-[(1E)-2-(2,4-dinitrophenyl)diazen-1-yl]-4-hydroxynaphthalene-2,7-disulfonate
Synonyms: disodium 3-[(1E)-2-(2,4-dinitrophenyl)diazen-1-yl]-4-hydroxynaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 542.36
Molecular Formula: C16H8N4Na2O11S2
InChi Key: XBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES: [Na+].[Na+].OC1=C2C=CC(=CC2=CC(=C1\N=N\C1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
Color 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)