accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOCH3

CAS RN 542-52-9

IUPAC Name: dibutyl carbonate
Synonyms: carbonic acid, dibutyl ester
Molecular Weight (g/mol): 174.24
Molecular Formula: C9H18O3
InChi Key: QLVWOKQMDLQXNN-UHFFFAOYSA-N
SMILES: CCCCOC(=O)OCCCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results