accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 54149-17-6

H3COOBr

CAS RN 54149-17-6

IUPAC Name: 1-bromo-2-(2-methoxyethoxy)ethane
Synonyms: 1-bromo-2-(2-methoxyethoxy)ethane
Molecular Weight (g/mol): 183.05
Molecular Formula: C5H11BrO2
InChi Key: HUXJXNSHCKHFIL-UHFFFAOYSA-N
SMILES: COCCOCCBr
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)