accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5405-79-8

IUPAC Name: 2,2-dimethylhexan-3-one
Synonyms: 3-hexanone, 2,2-dimethyl-
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: PYCHXHVFOZBVEY-UHFFFAOYSA-N
SMILES: CCCC(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results