accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5402-60-8

IUPAC Name: 2-(chloromethyl)-1,3-dimethylbenzene
Synonyms: 2-(chloromethyl)-1,3-dimethylbenzene
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: HPVRFWQMBYLJRL-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1CCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results