accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SnH3CCH3H3CH3C

CAS RN 53911-92-5

IUPAC Name: tributyl(3-methylbut-2-en-1-yl)stannane
Synonyms: tributyl(3-methylbut-2-en-1-yl)stannane
Molecular Weight (g/mol): 359.19
Molecular Formula: C17H36Sn
InChi Key: XEFRYQPTIWSVGI-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)CC=C(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)