accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5382-16-1

OHNH2

CAS RN 5382-16-1

IUPAC Name: 4-hydroxypiperidin-1-ium
Synonyms: 4-hydroxypiperidin-1-ium
Molecular Weight (g/mol): 102.16
Molecular Formula: C5H12NO
InChi Key: HDOWRFHMPULYOA-UHFFFAOYSA-O
SMILES: OC1CC[NH2+]CC1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)