accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 537-45-1

ClNBrOBr

CAS RN 537-45-1

IUPAC Name: 2,6-dibromo-4-(chloroimino)cyclohexa-2,5-dien-1-one
Synonyms: 2,6-dibromoquinonechloroimide
Molecular Weight (g/mol): 299.35
Molecular Formula: C6H2Br2ClNO
InChi Key: JYWKEVKEKOTYEX-UHFFFAOYSA-N
SMILES: ClN=C1C=C(Br)C(=O)C(Br)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)