accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 536693-97-7

BNClClHOOH

CAS RN 536693-97-7

IUPAC Name: (2,5-dichloropyridin-3-yl)boronic acid
Synonyms: 2,5-dichloropyridin-3-ylboronic acid
Molecular Weight (g/mol): 191.8
Molecular Formula: C5H4BCl2NO2
InChi Key: IISUHTJFLBUMKT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results