accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 53575-08-9

H3COPOOH3C

CAS RN 53575-08-9

IUPAC Name: diethyl [(naphthalen-1-yl)methyl]phosphonate
Synonyms: diethyl naphthalen-1-ylmethylphosphonate
Molecular Weight (g/mol): 278.29
Molecular Formula: C15H19O3P
InChi Key: AFNZPMQIKMSUTD-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=C2C=CC=CC2=CC=C1)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results