accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5345-27-7

IUPAC Name: 3-methanesulfonylbenzoate
Synonyms: 3-methanesulfonylbenzoate
Molecular Weight (g/mol): 199.2
Molecular Formula: C8H7O4S
InChi Key: KUTBMATZUQWFSR-UHFFFAOYSA-M
SMILES: CS(=O)(=O)C1=CC=CC(=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results