accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNONOOCl

CAS RN 5344-54-7

IUPAC Name: N-(3-chloro-4-methylphenyl)-4-nitrobenzamide
Synonyms: N-(3-chloro-4-methylphenyl)-4-nitrobenzamide
Molecular Weight (g/mol): 290.7
Molecular Formula: C14H11ClN2O3
InChi Key: WVMKIXPORUXIMJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(=O)C2=CC=C(C=C2)[N+]([O-])=O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results