accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 534-00-9

CH3(S)H3CBr

CAS RN 534-00-9

IUPAC Name: (2S)-1-bromo-2-methylbutane
Synonyms: (2S)-1-bromo-2-methylbutane
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: XKVLZBNEPALHIO-YFKPBYRVSA-N
SMILES: CC[C@H](C)CBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results