accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5336-08-3

IUPAC Name: 3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
Synonyms: ribonolactone
Molecular Weight (g/mol): 148.11
Molecular Formula: C5H8O5
InChi Key: CUOKHACJLGPRHD-UHFFFAOYNA-N
SMILES: OCC1OC(=O)C(O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)