accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5333-27-7

H3COCH3NOO

CAS RN 5333-27-7

IUPAC Name: 1-(4-methyl-3-nitrophenyl)ethan-1-one
Synonyms: 1-(4-methyl-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: YRBBMDOBWRUMEZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results