accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5331-43-1

IUPAC Name: (benzyloxy)carbohydrazide
Synonyms: benzyloxycarbohydrazide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: RXUBZLMIGSAPEJ-UHFFFAOYSA-N
SMILES: NNC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results