accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5329-33-9

HClH3CONH2HN

CAS RN 5329-33-9

IUPAC Name: hydrogen methoxymethanimidamide chloride
Synonyms: hydrogen methoxymethanimidamide chloride
Molecular Weight (g/mol): 110.54
Molecular Formula: C2H7ClN2O
InChi Key: MUDVUWOLBJRUGF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(N)=N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results