accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3BrNOO

CAS RN 5326-34-1

IUPAC Name: 1-bromo-4-methyl-2-nitrobenzene
Synonyms: 1-bromo-4-methyl-2-nitrobenzene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: UPBUTKQMDPHQAQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C(=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results