accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NONOH3CI(E/Z)(E/Z)(E/Z)

CAS RN 53213-92-6

IUPAC Name: 3-pentyl-2-[5-(3-pentyl-2,3-dihydro-1,3-benzoxazol-2-ylidene)penta-1,3-dien-1-yl]-1,3-benzoxazol-3-ium iodide
Synonyms: 3-pentyl-2-[5-(3-pentyl-1,3-benzoxazol-2-ylidene)penta-1,3-dien-1-yl]-1,3-benzoxazol-3-ium iodide
Molecular Weight (g/mol): 570.52
Molecular Formula: C29H35IN2O2
InChi Key: WSRFDTBIGBFQOK-UHFFFAOYSA-M
SMILES: [I-].CCCCCN1C(OC2=CC=CC=C12)=CC=CC=CC1=[N+](CCCCC)C2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results