accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 53137-27-2

CH3SOOH3CN

CAS RN 53137-27-2

IUPAC Name: dimethyl-1,3-thiazole-5-carboxylate
Synonyms: dimethyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 156.18
Molecular Formula: C6H6NO2S
InChi Key: MQGBARXPCXAFRZ-UHFFFAOYSA-M
SMILES: CC1=NC(C)=C(S1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results