accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 53087-78-8

H3COONBr

CAS RN 53087-78-8

IUPAC Name: ethyl 2-bromopyridine-3-carboxylate
Synonyms: ethyl 2-bromopyridine-3-carboxylate
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: QOFKYVXBHUEWBX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Br)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results