accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOSCH3

CAS RN 53053-51-3

IUPAC Name: methyl 4-(methylsulfanyl)butanoate
Synonyms: methyl 4-(methylsulfanyl)butanoate
Molecular Weight (g/mol): 148.22
Molecular Formula: C6H12O2S
InChi Key: HQDLZISNHUMXEA-UHFFFAOYSA-N
SMILES: COC(=O)CCCSC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)