CAS RN 5301-78-0
CAS RN 5301-78-0
IUPAC Name:
4-(hydroxymethyl)-2,6,7-trioxa-1λ⁵-phosphabicyclo[2.2.2]octan-1-one
Synonyms:
4-(hydroxymethyl)-2,6,7-trioxa-1λ⁵-phosphabicyclo[2.2.2]octan-1-one
Molecular Weight (g/mol):
180.1
Molecular Formula:
C5H9O5P
InChi Key:
YASRHLDAFCMIPB-UHFFFAOYSA-N
SMILES:
OCC12COP(=O)(OC1)OC2
1
–
1
of
1
results
1
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-oxide, 98%
CAS: 5301-78-0 Molecular Formula: C5H9O5P Molecular Weight (g/mol): 180.096 MDL Number: MFCD01677612 InChI Key: YASRHLDAFCMIPB-UHFFFAOYSA-N PubChem CID: 199942 IUPAC Name: (1-oxo-2,6,7-trioxa-1$l^{5}-phosphabicyclo[2.2.2]octan-4-yl)methanol SMILES: C1C2(COP(=O)(O1)OC2)CO