accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5296-64-0

IUPAC Name: (prop-2-en-1-ylsulfanyl)benzene
Synonyms: allyl phenyl sulfide
Molecular Weight (g/mol): 150.24
Molecular Formula: C9H10S
InChi Key: QGNRLAFFKKBSIM-UHFFFAOYSA-N
SMILES: C=CCSC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results