accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5296-62-8

IUPAC Name: 3-(ethylsulfanyl)prop-1-ene
Synonyms: 3-(ethylsulfanyl)prop-1-ene
Molecular Weight (g/mol): 102.2
Molecular Formula: C5H10S
InChi Key: NOJXPGXFDASWEI-UHFFFAOYSA-N
SMILES: CCSCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results