accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CPPH3CCH3H3CCH3H3CCH3

CAS RN 528854-34-4

IUPAC Name: 1-{2-[2,5-bis(propan-2-yl)phospholan-1-yl]ethyl}-2,5-bis(propan-2-yl)phospholane
Synonyms: 1-[2-(2,5-diisopropylphospholan-1-yl)ethyl]-2,5-diisopropylphospholane
Molecular Weight (g/mol): 370.54
Molecular Formula: C22H44P2
InChi Key: IRVIFEWWPYKALC-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C(C)C)P1CCP1C(CCC1C(C)C)C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results