accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 52671-45-1

ONHONONClClH2OH2ONa

CAS RN 52671-45-1

IUPAC Name: 1,3-dichloro-1,3,5-triazinane-2,4,6-trione dihydrate sodium
Synonyms: dichloroisocyanuric acid dihydrate sodium
Molecular Weight (g/mol): 256.98
Molecular Formula: C3H5Cl2N3NaO5
InChi Key: FIHQYHSNTSJNGG-UHFFFAOYSA-N
SMILES: O.O.[Na].ClN1C(=O)NC(=O)N(Cl)C1=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results