accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SN

CAS RN 5263-57-0

IUPAC Name: (cyclopentylsulfanyl)formonitrile
Synonyms: (cyclopentylsulfanyl)formonitrile
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: ZTZNTHBAUFZQGQ-UHFFFAOYSA-N
SMILES: N#CSC1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results