accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NaNN(R)NHNPO(R)(S)OOOOH2NOHONa

CAS RN 52558-16-4

IUPAC Name: [(2R,3S,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Synonyms: [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol): 345.21
Molecular Formula: C10H12N5O7P
InChi Key: LTFMZDNNPPEQNG-KVQBGUIXSA-L
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)[C@H]1C[C@H](O)[C@@H](COP([O-])([O-])=O)O1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results