accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOOClClClPdPd(E/Z)(E/Z)(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 52522-40-4

IUPAC Name: tris(1,5-diphenylpenta-1,4-dien-3-one) trichloromethane dipalladium
Synonyms: chloroform tris(dibenzylideneacetone) dipalladium
Molecular Weight (g/mol): 1035.1
Molecular Formula: C52H43Cl3O3Pd2
InChi Key: LNAMMBFJMYMQTO-UHFFFAOYSA-N
SMILES: [Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)