accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH(R)HNOHOOCH3CH3CH3O

CAS RN 5241-64-5

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 303.34
Molecular Formula: C16H19N2O4
InChi Key: NFVNYBJCJGKVQK-CYBMUJFWSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)