accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ON

CAS RN 52407-43-9

IUPAC Name: 2-(1-benzofuran-3-yl)acetonitrile
Synonyms: 2-(1-benzofuran-3-yl)acetonitrile
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: BJAJKVZABJZXDC-UHFFFAOYSA-N
SMILES: N#CCC1=COC2=CC=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results