accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 52364-71-3

IUPAC Name: 4'-(pentyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(pentyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 265.36
Molecular Formula: C18H19NO
InChi Key: RDISTOCQRJJICR-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results