accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONH2HN

CAS RN 52328-05-9

IUPAC Name: methoxymethanimidamide
Synonyms: methoxymethanimidamide
Molecular Weight (g/mol): 74.08
Molecular Formula: C2H6N2O
InChi Key: RMAHPRNLQIRHIJ-UHFFFAOYSA-N
SMILES: COC(N)=N
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)