accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5231-87-8

CH3OOCH3OO

CAS RN 5231-87-8

IUPAC Name: diethoxycyclobut-3-ene-1,2-dione
Synonyms: diethoxycyclobut-3-ene-1,2-dione
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: DFSFLZCLKYZYRD-UHFFFAOYSA-N
SMILES: CCOC1=C(OCC)C(=O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)