accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 52253-69-7

HH2OBrNH2SN

CAS RN 52253-69-7

IUPAC Name: hydrogen 4-phenyl-1,3-thiazol-2-amine hydrate bromide
Synonyms: hydrogen phenthiazamine hydrate bromide
Molecular Weight (g/mol): 275.16
Molecular Formula: C9H11BrN2OS
InChi Key: SBVYKOGRUCDICV-UHFFFAOYSA-N
SMILES: [H+].O.[Br-].NC1=NC(=CS1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results