accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3OO

CAS RN 5222-73-1

IUPAC Name: dimethoxycyclobut-3-ene-1,2-dione
Synonyms: dimethoxycyclobut-3-ene-1,2-dione
Molecular Weight (g/mol): 142.11
Molecular Formula: C6H6O4
InChi Key: SZBNZTGCAMLMJY-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results