accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5220-49-5

NH2O

CAS RN 5220-49-5

IUPAC Name: 3-aminocyclohex-2-en-1-one
Synonyms: 3-aminocyclohex-2-en-1-one
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: ZZMRPOAHZITKBV-UHFFFAOYSA-N
SMILES: NC1=CC(=O)CCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)