CAS RN 52080-58-7
IUPAC Name:
4-{[4-(dimethylamino)phenyl][4-(methylimino)cyclohexa-2,5-dien-1-ylidene]methyl}-N,N-dimethylaniline
Synonyms:
4-{[4-(dimethylamino)phenyl][4-(methylimino)cyclohexa-2,5-dien-1-ylidene]methyl}-N,N-dimethylaniline
Molecular Weight (g/mol):
357.5
Molecular Formula:
C24H27N3
InChi Key:
AMPCGOAFZFKBGH-UHFFFAOYSA-N
SMILES:
CN=C1C=CC(C=C1)=C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
1
–
1
of
1
results
1
Methyl Violet (Technical Grade), TRC
CAS: 52080-58-7 Molecular Formula: C24 H27 N3 Molecular Weight (g/mol): 357.491 Synonym: C.I. Solvent Violet 8,Aizen Methyl Violet Base,Baso Violet 608,Benzenamine, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-, N-Me derivs.,Brilliant Oil Violet Base,C.I. 42535:1,C.I. 42535B,Fat Soluble Violet K,Methyl Violet 2B Base IUPAC Name: 4-[[4-(dimethylamino)phenyl]-(4-methyliminocyclohexa-2,5-dien-1-ylidene)methyl]-N,N-dimethylaniline SMILES: CN=C1C=CC(=C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C)C=C1