accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 52067-36-4

CH3OCH3HO

CAS RN 52067-36-4

IUPAC Name: 1-(4-ethoxyphenyl)ethan-1-ol
Synonyms: 1-(4-ethoxyphenyl)ethanol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: GKGQWBJLOYXULB-UHFFFAOYNA-N
SMILES: CCOC1=CC=C(C=C1)C(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results