accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5182-44-5

OHCl

CAS RN 5182-44-5

IUPAC Name: 2-(3-chlorophenyl)ethan-1-ol
Synonyms: 2-(3-chlorophenyl)ethanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: NDWAVJKRSASRPH-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC(Cl)=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)