accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNON

CAS RN 51791-12-9

IUPAC Name: 3-(chloromethyl)-1,2,4-oxadiazole
Synonyms: 3-(chloromethyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 118.52
Molecular Formula: C3H3ClN2O
InChi Key: YSNKGJCEHOJIDK-UHFFFAOYSA-N
SMILES: ClCC1=NOC=N1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results